missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Dimethylaminoethyl)pyridine, MP Biomedicals

Product Code. 11472835 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 11472835

Brand: MP Biomedicals 0521103510

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 6304-27-4
Molecular Formula C9H14N2
MDL Number MFCD00023546
Quantity 10 g
Synonym 2-2-dimethylaminoethyl pyridine, pyridine, 2-2-dimethylamino ethyl, 2-2-dimethylamino ethyl pyridine, dimethyl 2-pyridin-2-yl ethyl amine, n,n-dimethylpyridine-2-ethylamine, 2-pyridineethanamine, n,n-dimethyl, 2-beta-dimethylamino ethyl pyridine, 2-2-dimethylamino-ethyl-pyridine, n,n-dimethyl-n-2-2-pyridyl ethyl amine
InChI Key FXHJGPDCPMCUKW-UHFFFAOYSA-N
SMILES CN(C)CCC1=CC=CC=N1
IUPAC Name dimethyl[2-(pyridin-2-yl)ethyl]amine
Molecular Weight (g/mol) 150.23
PubChem CID 80554
Formula Weight 150.23
Chemical Name or Material 2-(2-Dimethylaminoethyl)pyridine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.