accessibility menu, dialog, popup

UserName

CAS RN 62208-63-3

CH3NNNOO

CAS RN 62208-63-3

IUPAC Name: 1-methyl-4-(2-nitrophenyl)piperazine
Synonyms: 1-methyl-4-(2-nitrophenyl)piperazine
Molecular Weight (g/mol): 221.26
Molecular Formula: C11H15N3O2
InChi Key: SSRLRVMFEGGGMQ-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1=CC=CC=C1[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results