CAS RN 61438-64-0
CAS RN 61438-64-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Closantel sodium, MedChemExpress
MedChemExpress Closantel sodium is a halogenated salicylanilide with a potent anti-parasitic activity. Closantel sodium is a potent and highly specific Onchocerca volvulus chitinase (OvCHT1) inhibitor with an IC50 of 1.6 μM and a Ki of 468 nM. Closantel sodium inhibits the O. volvulus L3 to L4 molt of developing.