Learn More
Closantel sodium, MedChemExpress
Product Code. 16477386
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
Product Code. 16477386
Brand: Medchem Express HY17596A
MedChemExpress Closantel sodium is a halogenated salicylanilide with a potent anti-parasitic activity. Closantel sodium is a potent and highly specific Onchocerca volvulus chitinase (OvCHT1) inhibitor with an IC50 of 1.6 μM and a Ki of 468 nM. Closantel sodium inhibits the O. volvulus L3 to L4 molt of developing.
Closantel
Specifications
Chemical Name or Material Closantel sodium
Physical Form Solid
CAS 61438-64-0
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C22H13Cl2I2N2NaO2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 125 mg/mL (182.47 mM; Need ultrasonic)
SMILES O=C(NC1=CC(Cl)=C(C(C2=CC=C(Cl)C=C2)C#N)C=C1C)C3=CC(I)=CC(I)=C3O[Na]
Molecular Weight (g/mol) 685.06
Formula Weight 685.06
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.