CAS RN 61422-45-5
CAS RN 61422-45-5
IUPAC Name:
5-fluoro-N-hexyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-1-carboxamide
Synonyms:
carmofur 卡莫氟
Molecular Weight (g/mol):
257.27
Molecular Formula:
C11H16FN3O3
InChi Key:
AOCCBINRVIKJHY-UHFFFAOYSA-N
SMILES:
CCCCCCNC(=O)N1C=C(F)C(=O)NC1=O
1
–
1
of
1
results
1
Carmofur, MedChemExpress
MedChemExpress Carmofur (HCFU), a derivative of 5-Fluorouracil, is an antineoplastic agent. Carmofur is an inhibitor of acid ceramidase with an IC50 of 79 nM for the rat enzyme. Carmofur inhibits the SARS-CoV-2 main protease (Mpro). Carmofur Inhibits SARS-CoV-2 in Vero E6 cell with an EC50 of 24.3 μM.