Learn More
Carmofur, MedChemExpress
Product Code. 16478156
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. 16478156
Brand: Medchem Express HYB0182
MedChemExpress Carmofur (HCFU), a derivative of 5-Fluorouracil, is an antineoplastic agent. Carmofur is an inhibitor of acid ceramidase with an IC50 of 79 nM for the rat enzyme. Carmofur inhibits the SARS-CoV-2 main protease (Mpro). Carmofur Inhibits SARS-CoV-2 in Vero E6 cell with an EC50 of 24.3 μM.
Carmofur (HCFU), a derivative of 5-Fluorouracil, is an antineoplastic agent. Carmofur is an inhibitor of acid ceramidase with an IC50 of 79 nM for the rat enzyme. Carmofur inhibits the SARS-CoV-2 main protease (Mpro). Carmofur Inhibits SARS-CoV-2 in Vero E6 cell with an EC50 of 24.3 μM.
Specifications
Chemical Name or Material Carmofur
Physical Form Solid
CAS 61422-45-5
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C11H16FN3O3
For Use With (Application) Cancer-programmed cell death
Synonym HCFU
Solubility Information DMSO : ≥ 100 mg/mL (388.71 mM) ∣H2O : 0.67 mg/mL (2.60 mM; Need ultrasonic)
SMILES O=C(N(C(N1)=O)C=C(F)C1=O)NCCCCCC
Molecular Weight (g/mol) 257.26
Formula Weight 257.26
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.