accessibility menu, dialog, popup

UserName

CAS RN 6106-22-5

NH2SOOOH2OH2ONa

CAS RN 6106-22-5

IUPAC Name: sodium 4-aminobenzene-1-sulfonate dihydrate
Synonyms: sodium 4-aminobenzenesulfonate dihydrate
Molecular Weight (g/mol): 231.2
Molecular Formula: C6H10NNaO5S
InChi Key: MJFZIDUBUNIFLZ-UHFFFAOYSA-M
SMILES: O.O.[Na+].NC1=CC=C(C=C1)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results