missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfanilate Dihydrate, Sigma-Aldrich™
Specifications
Specifications
| CAS | 6106-22-5 |
| Quantity | 250 g |
| Molecular Formula | 4-(NH2)C6H4SO3Na · 2H2O |
| MDL Number | MFCD00151042 |
| Synonym | Sodium sulfanilate dihydrate, Sulfanilic acid sodium salt, Sodium sulfanilate dihydrate |
| Molecular Weight (g/mol) | 231.2 |
| Chemical Name or Material | Sodium Sulfanilate Dihydrate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?