accessibility menu, dialog, popup

UserName

OSOOHNOOHCl

CAS RN 606944-48-3

IUPAC Name: 4-[4-(2-chlorophenoxy)benzenesulfonamido]benzoic acid
Synonyms: 4-[4-(2-chlorophenoxy)benzenesulfonamido]benzoic acid
Molecular Weight (g/mol): 403.83
Molecular Formula: C19H14ClNO5S
InChi Key: OMOBIVNVBIPQNK-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(NS(=O)(=O)C2=CC=C(OC3=CC=CC=C3Cl)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results