CAS RN 600734-06-3
CAS RN 600734-06-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bay 65-1942 hydrochloride, MedChemExpress
MedChemExpress Bay 65-1942 hydrochloride is an ATP-competitive and selective IKKβ inhibitor.