Learn More
Bay 65-1942 hydrochloride, MedChemExpress
Product Code. 16440608
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
Unit Size:
10mg
2mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16440608 5 mg 5mg
16440356 2 mg 2mg
16418158 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16440608 Supplier Medchem Express Supplier No. HY50948
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
MedChemExpress Bay 65-1942 hydrochloride is an ATP-competitive and selective IKKβ inhibitor.
Bay 65-1942 hydrochloride is an ATP-competitive and selective IKKβ inhibitor.
Specifications
Chemical Name or Material Bay 65-1942 hydrochloride
CAS 600734-06-3
Color White
Purity Grade Notes Research
Molecular Formula C22H26ClN3O4
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (115.76 mM; Need ultrasonic) ∣H2O : 2.17 mg/mL (5.02 mM; Need ultrasonic)
SMILES O=C1OCC2=C([C@H]3CNCCC3)C=C(C4=C(O)C=CC=C4OCC5CC5)N=C2N1.[H]Cl
Molecular Weight (g/mol) 431.91
Formula Weight 431.91
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation