accessibility menu, dialog, popup

UserName

CH3H2NOCH3OCH3OH3C

CAS RN 58780-82-8

IUPAC Name: methyl[(3,4,5-trimethoxyphenyl)methyl]azanium
Synonyms: methyl[(3,4,5-trimethoxyphenyl)methyl]azanium
Molecular Weight (g/mol): 212.27
Molecular Formula: C11H18NO3
InChi Key: LFULMNRPABTDDQ-UHFFFAOYSA-O
SMILES: C[NH2+]CC1=CC(OC)=C(OC)C(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results