accessibility menu, dialog, popup

UserName

NH2SNHNCl(E)

CAS RN 5706-78-5

IUPAC Name: [(E)-[(2-chlorophenyl)methylidene]amino]thiourea
Synonyms: (E)-[(2-chlorophenyl)methylidene]aminothiourea
Molecular Weight (g/mol): 213.68
Molecular Formula: C8H8ClN3S
InChi Key: ZGPJVXICCFYSRT-VZUCSPMQSA-N
SMILES: NC(=S)N\N=C\C1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results