accessibility menu, dialog, popup

UserName

H3CNHOONHCH3(R)NHCH3(S)H3COOO

CAS RN 57-64-7

IUPAC Name: (3aS,8aR)-1,3a,8-trimethyl-5-[(methylcarbamoyl)oxy]-1H,2H,3H,3aH,8H,8aH-pyrrolo[2,3-b]indole-1,8-diium 2-oxidobenzoate
Synonyms: (3aS,8aR)-1,3a,8-trimethyl-5-[(methylcarbamoyl)oxy]-1H,2H,3H,8H,8aH-pyrrolo[2,3-b]indole-1,8-diium 2-oxidobenzoate
Molecular Weight (g/mol): 413.47
Molecular Formula: C22H27N3O5
InChi Key: HZOTZTANVBDFOF-PBCQUBLHSA-N
SMILES: [O-]C(=O)C1=CC=CC=C1[O-].CNC(=O)OC1=CC=C2[NH+](C)[C@H]3[NH+](C)CC[C@@]3(C)C2=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results