missing translation for 'onlineSavingsMsg'
Learn More

Eserine Salicylate, MP Biomedicals

Brand:  MP Biomedicals 0521376701

Quantity 1 g

Product Code. 11443135

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

  • Important Notice: This product belongs to MP Biomedicals Rare Chemical library
  • Most Rare Chemical products are available for immediate delivery and sold 'As Is'
  • MP Biomedicals does not collect analytical data for those products
  • However, analytical test may be performed upon custom request for bulk orders

TRUSTED_SUSTAINABILITY
Specifications

Specifications

57-64-7
C22H27N3O5
MFCD00135659
physostigmine salicylate, unii-2046zro9vu, antilirium tn, 3ar,8bs-3,4,8b-trimethyl-2,3a-dihydro-1h-pyrrolo 2,3-b indol-7-yl n-methylcarbamate;2-hydroxybenzoic acid, physostigmine salicylate usp:jan, spectrum_000223, opera_id_194, physostigmine salicylate salt, physostigmine; salicyclic acid, spectrum1500486
CC12CCN(C1N(C3=C2C=C(C=C3)OC(=O)NC)C)C.C1=CC=C(C(=C1)C(=O)O)O
413.474
CHEBI:48883
ESERINE SALICYLATE
181°C to 183°C (literature)
C15H21N3O2 . C7H6O3
1 g
HZOTZTANVBDFOF-PBCQUBLHSA-N
[(3aR,8bS)-3,4,8b-trimethyl-2,3a-dihydro-1H-pyrrolo[2,3-b]indol-7-yl] N-methylcarbamate;2-hydroxybenzoic acid
657348
413.45
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.