accessibility menu, dialog, popup

UserName

CAS RN 5521-56-2

IUPAC Name: 6-methylpyrazin-2-amine
Synonyms: pyrazinamine, 6-methyl-
Molecular Weight (g/mol): 109.13
Molecular Formula: C5H7N3
InChi Key: UAOOJJPSCLNTOP-UHFFFAOYSA-N
SMILES: CC1=CN=CC(N)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results