accessibility menu, dialog, popup

UserName

H3COOHONH2F

CAS RN 54788-29-3

IUPAC Name: 2-amino-3-(2-fluoro-4-methoxyphenyl)propanoic acid
Synonyms: 2-amino-3-(2-fluoro-4-methoxyphenyl)propanoic acid
Molecular Weight (g/mol): 213.21
Molecular Formula: C10H12FNO3
InChi Key: WPRRDCRYBYGCQS-UHFFFAOYNA-N
SMILES: COC1=CC(F)=C(CC(N)C(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)