accessibility menu, dialog, popup

UserName

NHN

CAS RN 5379-94-2

IUPAC Name: 3-[(pyrrolidin-1-yl)methyl]-1H-indole
Synonyms: 3-(pyrrolidin-1-ylmethyl)-1H-indole
Molecular Weight (g/mol): 200.29
Molecular Formula: C13H16N2
InChi Key: ZIIMZMSKBKAABD-UHFFFAOYSA-N
SMILES: C(N1CCCC1)C1=CNC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results