CAS RN 52364-72-4
IUPAC Name:
4'-(heptyloxy)-[1,1'-biphenyl]-4-carbonitrile
Synonyms:
4'-(heptyloxy)-[1,1'-biphenyl]-4-carbonitrile
Molecular Weight (g/mol):
293.41
Molecular Formula:
C20H23NO
InChi Key:
JPBFKTCKZLMJED-UHFFFAOYSA-N
SMILES:
CCCCCCCOC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
1
–
1
of
1
results
1
4-Cyano-4'-n-heptyloxybiphenyl, 99%, Thermo Scientific™
CAS: 52364-72-4 Molecular Formula: C20H23NO Molecular Weight (g/mol): 293.41 MDL Number: MFCD00134146 InChI Key: JPBFKTCKZLMJED-UHFFFAOYSA-N Synonym: 4'-heptyloxy-4-cyanobiphenyl,4-cyano-4'-heptyloxybiphenyl,4'-heptyloxy-1,1'-biphenyl-4-carbonitrile,4'-heptyloxy biphenyl-4-carbonitrile,7cob,1,1'-biphenyl-4-carbonitrile, 4'-heptyloxy,4-heptyloxycyanodiphenyl,4-cyano-4'-heptoxybiphenyl,4-4-heptoxyphenyl benzonitrile,4-4-heptyloxyphenyl benzonitrile PubChem CID: 104172 IUPAC Name: 4-(4-heptoxyphenyl)benzonitrile SMILES: CCCCCCCOC1=CC=C(C=C1)C2=CC=C(C=C2)C#N