CAS RN 51904-91-7
IUPAC Name:
5-(chlorosulfonyl)-2-methoxybenzoic acid
Synonyms:
5-(chlorosulfonyl)-2-methoxybenzoic acid
Molecular Weight (g/mol):
250.65
Molecular Formula:
C8H7ClO5S
InChi Key:
OUBZCDOQEMLMAB-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1C(O)=O)S(Cl)(=O)=O
1
–
1
of
1
results
1
5-Chlorosulfonyl-2-methoxybenzoic acid, 99%, Thermo Scientific™
CAS: 51904-91-7 Molecular Formula: C8H7ClO5S Molecular Weight (g/mol): 250.649 MDL Number: MFCD00066912 InChI Key: OUBZCDOQEMLMAB-UHFFFAOYSA-N Synonym: 5-chlorosulfonyl-2-methoxybenzoic acid,5-chlorosulphonyl-2-anisic acid,benzoic acid, 5-chlorosulfonyl-2-methoxy,5-chlorosulphonyl-2-methoxybenzoic acid,5-chlorosulfonyl-2-methoxybenzoicacid,acmc-1ar14,2-methoxy-5-chlorosulfonylbenzoic acid,5-chlorosulfonyl-2-methoxy-benzoic acid,5-chlorosulphonyl-2-methoxy benzoic acid,5-chlorosulphonyl-2-methoxy-benzoic acid PubChem CID: 104007 IUPAC Name: 5-chlorosulfonyl-2-methoxybenzoic acid SMILES: COC1=C(C=C(C=C1)S(=O)(=O)Cl)C(=O)O