accessibility menu, dialog, popup

UserName

CAS RN 50402-56-7

H2NNHSOHOO

CAS RN 50402-56-7

IUPAC Name: 5-[(2-aminoethyl)amino]naphthalene-1-sulfonic acid
Synonyms: edans
Molecular Weight (g/mol): 266.32
Molecular Formula: C12H14N2O3S
InChi Key: SJQRQOKXQKVJGJ-UHFFFAOYSA-N
SMILES: NCCNC1=C2C=CC=C(C2=CC=C1)S(O)(=O)=O
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)