Learn More
N-(Aminoethyl)-5-naphthylamine-1-sulfonic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | N-(Aminoethyl)-5-naphthylamine-1-sulfonic Acid |
| CAS | 50402-56-7 |
| Molecular Formula | C12 H14 N2 O3 S |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C12H14N2O3S/c13-7-8-14-11-5-1-4-10-9(11)3-2-6-12(10)18(15,16)17/h1-6,14H,7-8,13H2,(H,15,16,17) |
| SMILES | NCCNc1cccc2c(cccc12)S(=O)(=O)O |
| IUPAC Name | 5-(2-aminoethylamino)naphthalene-1-sulfonic acid |
| Molecular Weight (g/mol) | 266.32 |
| Formula Weight | 266.0725 |