accessibility menu, dialog, popup

UserName

CAS RN 438553-44-7

IUPAC Name: potassium trifluoro(3-methoxyphenyl)boranuide
Synonyms: potassium trifluoro(3-methoxyphenyl)boranuide
Molecular Weight (g/mol): 214.04
Molecular Formula: C7H7BF3KO
InChi Key: GKDCWEPJOSDGHY-UHFFFAOYSA-N
SMILES: [K+].COC1=CC=CC(=C1)[B-](F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results