accessibility menu, dialog, popup

UserName

CAS RN 4248-33-3

IUPAC Name: 3,4-dinitrobenzonitrile
Synonyms: 3,4-dinitrobenzonitrile
Molecular Weight (g/mol): 193.12
Molecular Formula: C7H3N3O4
InChi Key: QXFXDEHCVXHEBX-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1[N+]([O-])=O)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results