CAS RN 37798-08-6
IUPAC Name:
1-(1-benzofuran-5-yl)methanamine
Synonyms:
1-(1-benzofuran-5-yl)methanamine
Molecular Weight (g/mol):
147.18
Molecular Formula:
C9H9NO
InChi Key:
OFMIMSPBHXZKRI-UHFFFAOYSA-N
SMILES:
NCC1=CC=C2OC=CC2=C1
1
–
1
of
1
results
1
1-Benzofuran-5-ylmethylamine, 97%, Thermo Scientific™
CAS: 37798-08-6 Molecular Formula: C9H9NO Molecular Weight (g/mol): 147.177 MDL Number: MFCD04974039 InChI Key: OFMIMSPBHXZKRI-UHFFFAOYSA-N Synonym: 1-benzofuran-5-ylmethylamine,benzofuran-5-ylmethanamine,5-aminomethyl benzofuran,1-1-benzofuran-5-yl methanamine,pubchem7027,5-benzofuranylmethanamine,benzo b furan-5-ylmethylamine,c-benzofuran-5-yl-methylamine,1-benzofuran-5-yl methanamine,1-benzo b furan-5-ylmethylamine PubChem CID: 2795182 IUPAC Name: 1-benzofuran-5-ylmethanamine SMILES: C1=CC2=C(C=CO2)C=C1CN