accessibility menu, dialog, popup

UserName

CAS RN 374537-99-2

HNBrNH2

CAS RN 374537-99-2

IUPAC Name: 5-bromo-1H-indol-7-amine
Synonyms: 5-bromo-1H-indol-7-amine
Molecular Weight (g/mol): 211.06
Molecular Formula: C8H7BrN2
InChi Key: ZRTCJQGCGWZTAG-UHFFFAOYSA-N
SMILES: NC1=C2NC=CC2=CC(Br)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results