accessibility menu, dialog, popup

UserName

NOCl

CAS RN 349089-31-2

IUPAC Name: 4-chloro-N,N-dicyclohexylbenzamide
Synonyms: 4-chloro-N,N-dicyclohexylbenzamide
Molecular Weight (g/mol): 319.87
Molecular Formula: C19H26ClNO
InChi Key: BMQOLGWVHWPNTP-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C(=O)N(C1CCCCC1)C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results