accessibility menu, dialog, popup

UserName

CAS RN 261762-45-2

IUPAC Name: 1-(2-chloro-3,6-difluorophenyl)methanamine
Synonyms: 1-(2-chloro-3,6-difluorophenyl)methanamine
Molecular Weight (g/mol): 177.58
Molecular Formula: C7H6ClF2N
InChi Key: KDAQEMQCJMFSEP-UHFFFAOYSA-N
SMILES: NCC1=C(F)C=CC(F)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results