accessibility menu, dialog, popup

UserName

CH3SSNH2NN

CAS RN 25660-70-2

IUPAC Name: 5-(ethylsulfanyl)-1,3,4-thiadiazol-2-amine
Synonyms: 5-(ethylsulfanyl)-1,3,4-thiadiazol-2-amine
Molecular Weight (g/mol): 161.24
Molecular Formula: C4H7N3S2
InChi Key: VWRHSNKTSSIMGE-UHFFFAOYSA-N
SMILES: CCSC1=NN=C(N)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results