accessibility menu, dialog, popup

UserName

CAS RN 2451-01-6

H2OCH3CH3HOH3COH

CAS RN 2451-01-6

IUPAC Name: 4-(2-hydroxypropan-2-yl)-1-methylcyclohexan-1-ol hydrate
Synonyms: terpin hydrate (cis-form)
Molecular Weight (g/mol): 190.28
Molecular Formula: C10H22O3
InChi Key: JGKJMBOJWVAMIJ-UHFFFAOYSA-N
SMILES: O.CC(C)(O)C1CCC(C)(O)CC1
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results