Learn More
Terpin Hydrate, TRC
Specifications
Specifications
| Chemical Name or Material | Terpin Hydrate |
| CAS | 2451-01-6 |
| Molecular Formula | C10 H20 O2 . H2O |
| Quantity | 500 mg |
| Synonym | cis-4-Hydroxy-alpha,alpha,4-trimethylcyclohexanemethanol monohydrate, p-Menthane-1,8-diol monohydrate, Terpin hydrate, Terpin monohydrate, Terpinene hydrate, Terpinol hydrate, cis-Terpin hydrate, cis-p-Methane-1,8-diol monohydrate, cis-p-Menthane-1,8-diol monohydrate |
| InChI Formula | InChI=1S/C10H20O2.H2O/c1-9(2,11)8-4-6-10(3,12)7-5-8;/h8,11-12H,4-7H2,1-3H3;1H2/t8-,10+; |
| SMILES | O.CC(C)(O)[C@@H]1CC[C@](C)(O)CC1 |
| IUPAC Name | 4-(2-hydroxypropan-2-yl)-1-methylcyclohexan-1-ol;hydrate |
| Molecular Weight (g/mol) | 190.28 |
| Formula Weight | 190.1569 |