accessibility menu, dialog, popup

UserName

CAS RN 24167-56-4

CH3NH3COF

CAS RN 24167-56-4

IUPAC Name: 4-fluoro-N,N-dimethylbenzamide
Synonyms: 4-fluoro-N,N-dimethylbenzamide
Molecular Weight (g/mol): 167.18
Molecular Formula: C9H10FNO
InChi Key: NUOGEPIJFRZXIN-UHFFFAOYSA-N
SMILES: CN(C)C(=O)C1=CC=C(F)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results