CAS RN 22948-02-3
IUPAC Name:
(3-aminophenyl)sulfanide
Synonyms:
(3-aminophenyl)sulfanide
Molecular Weight (g/mol):
124.18
Molecular Formula:
C6H6NS
InChi Key:
KFFUEVDMVNIOHA-UHFFFAOYSA-M
SMILES:
NC1=CC=CC([S-])=C1
1
–
2
of
2
results
1
3-Aminothiophenol, 97%
CAS: 22948-02-3 Molecular Formula: C6H6NS Molecular Weight (g/mol): 124.18 MDL Number: MFCD00007791 InChI Key: KFFUEVDMVNIOHA-UHFFFAOYSA-M Synonym: 3-aminothiophenol,benzenethiol, 3-amino,3-mercaptoaniline,m-amino thiophenol,3-aminobenzene-1-thiol,3-amino thiophenol,3-amino-benzenethiol,pubchem2470,acmc-1co9c PubChem CID: 31577 SMILES: NC1=CC=CC([S-])=C1