accessibility menu, dialog, popup

UserName

ClClNCSCl

CAS RN 22134-07-2

IUPAC Name: 1,3,5-trichloro-2-isothiocyanatobenzene
Synonyms: 1,3,5-trichloro-2-isothiocyanatobenzene
Molecular Weight (g/mol): 238.51
Molecular Formula: C7H2Cl3NS
InChi Key: DSXVZIOQROMOEF-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(N=C=S)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results