CAS RN 2166487-21-2
CAS RN 2166487-21-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
SIRT5 inhibitor 1, MedChemExpress
MedChemExpress SIRT5 inhibitor 1 is a potent Human Sirtuin 5 deacylase inhibitor, with an IC50 of 0.11 μM.