accessibility menu, dialog, popup

UserName

CAS RN 2154-67-8

CH3H3COOCH3H3CNHO

CAS RN 2154-67-8

IUPAC Name: 1-hydroxy-2,2,5,5-tetramethyl-2,5-dihydro-1H-pyrrole-3-carboxylate
Synonyms: 1-hydroxy-2,2,5,5-tetramethylpyrrole-3-carboxylate
Molecular Weight (g/mol): 184.22
Molecular Formula: C9H14NO3
InChi Key: SADYUZSTFFUBBP-UHFFFAOYSA-M
SMILES: CC1(C)C=C(C([O-])=O)C(C)(C)N1O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results