CAS RN 2123489-30-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
WNK-IN-11, MedChemExpress
MedChemExpress WNK-IN-11 is an allosteric With-No-Lysine (WNK) kinase inhibitor, with an IC50 of 4 nM for WNK1.