Learn More
WNK-IN-11, MedChemExpress
Description
WNK-IN-11 is an allosteric With-No-Lysine (WNK) kinase inhibitor, with an IC50 of 4 nM for WNK1.
Specifications
Specifications
| Chemical Name or Material | WNK-IN-11 |
| CAS | 2123489-30-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H21Cl2N5OS |
| Quantity | 5 mg |
| Solubility Information | DMSO : 150 mg/mL (324.39 mM; Need ultrasonic) |
| SMILES | O=C(C1=CC(C2=CSC(NC)=N2)=NC=C1Cl)N3CCN(CC4=CC=C(Cl)C=C4)CC3 |
| Molecular Weight (g/mol) | 462.4 |
| Formula Weight | 462.4 |
| Show More |