CAS RN 207-08-9
CAS RN 207-08-9
IUPAC Name:
benzo[k]fluoranthene
Synonyms:
benzo[k]fluoranthene benzo(K)fluoranthene dibenzo(b,jk)fluorene 11,12-benzo(k)fluoranthene benzo(k)fluoranthene ccris 74 2,3,1',8'-binaphthylene 8,9-benzofluoranthene 8,9-benzfluoranthene 11,12-benzofluoranthene
Molecular Weight (g/mol):
252.32
Molecular Formula:
C20H12
InChi Key:
HAXBIWFMXWRORI-UHFFFAOYSA-N
SMILES:
C1=CC=C2C=C3C(=CC2=C1)C1=CC=CC2=CC=CC3=C12
1
–
2
of
2
results
1
PAH Mixture 931 10 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality