Learn More
PAH Mixture 931 10 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| CAS | 207-08-9, 50-32-8, 205-99-2, 191-24-2, 206-44-0, 193-39-5 |
| Quantity | 1 mL |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |