accessibility menu, dialog, popup

UserName

CAS RN 204327-96-8

N(S)NH

CAS RN 204327-96-8

IUPAC Name: (3S)-1,3-dibenzylpiperazine
Synonyms: (3S)-1,3-dibenzylpiperazine
Molecular Weight (g/mol): 266.39
Molecular Formula: C18H22N2
InChi Key: AJNDRRBUHVZNKU-SFHVURJKSA-N
SMILES: C([C@H]1CN(CC2=CC=CC=C2)CCN1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results