CAS RN 202982-66-9
IUPAC Name:
(2E)-3-(4-chloro-3-fluorophenyl)prop-2-enoic acid
Synonyms:
(2E)-3-(4-chloro-3-fluorophenyl)prop-2-enoic acid
Molecular Weight (g/mol):
200.59
Molecular Formula:
C9H6ClFO2
InChi Key:
MNELKRQRBRYHBV-DUXPYHPUSA-N
SMILES:
OC(=O)\C=C\C1=CC=C(Cl)C(F)=C1
1
–
1
of
1
results
1
4-Chloro-3-fluorocinnamic acid, 95%, Thermo Scientific™
CAS: 202982-66-9 Molecular Formula: C9H6ClFO2 Molecular Weight (g/mol): 200.593 MDL Number: MFCD00143294 InChI Key: MNELKRQRBRYHBV-DUXPYHPUSA-N Synonym: 4-chloro-3-fluorocinnamic acid,e-3-4-chloro-3-fluorophenyl acrylic acid,2e-3-4-chloro-3-fluorophenyl prop-2-enoic acid,3-4-chloro-3-fluoro-phenyl-acrylic acid,rarechem bk hw 0060,e-4-chloro-3-fluorocinnamic acid,3-4-chloro-3-fluorophenyl acrylic acid,e-3-4-chloro-3-fluorophenyl prop-2-enoic acid PubChem CID: 5708403 IUPAC Name: (E)-3-(4-chloro-3-fluorophenyl)prop-2-enoic acid SMILES: C1=CC(=C(C=C1C=CC(=O)O)F)Cl