accessibility menu, dialog, popup

UserName

CAS RN 192944-51-7

CH3OOHNN

CAS RN 192944-51-7

IUPAC Name: ethyl 1H-indazole-5-carboxylate
Synonyms: ethyl 1H-indazole-5-carboxylate
Molecular Weight (g/mol): 190.2
Molecular Formula: C10H10N2O2
InChi Key: SKABXDPLIJIWLR-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C2NN=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results