accessibility menu, dialog, popup

UserName

CH3HNOCl

CAS RN 18370-10-0

IUPAC Name: 3-chloro-N-methylbenzamide
Synonyms: 3-chloro-N-methylbenzamide
Molecular Weight (g/mol): 169.61
Molecular Formula: C8H8ClNO
InChi Key: WJJLGPQRUCWHKZ-UHFFFAOYSA-N
SMILES: CNC(=O)C1=CC(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results