accessibility menu, dialog, popup

UserName

CAS RN 1823301-50-3

NOONOOOO

CAS RN 1823301-50-3

IUPAC Name: 4-(2,5-dioxopyrrolidin-1-yl) 1-(9H-fluoren-9-yl)methyl piperidine-1,4-dicarboxylate
Synonyms: 4-(2,5-dioxopyrrolidin-1-yl) 1-(9H-fluoren-9-ylmethyl) piperidine-1,4-dicarboxylate
Molecular Weight (g/mol): 448.48
Molecular Formula: C25H24N2O6
InChi Key: RDBHMYCVGMHUQA-UHFFFAOYSA-N
SMILES: O=C(ON1C(=O)CCC1=O)C1CCN(CC1)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results