CAS RN 1821-52-9
CAS RN 1821-52-9
IUPAC Name:
2-hydroxy-3-(1H-indol-3-yl)propanoic acid
Synonyms:
indole-3-lactic acid 5-iHIPA indolelactate indole-3-lactate indolelactic acid
Molecular Weight (g/mol):
205.21
Molecular Formula:
C11H11NO3
InChi Key:
XGILAAMKEQUXLS-UHFFFAOYNA-N
SMILES:
OC(CC1=CNC2=CC=CC=C12)C(O)=O
1
–
1
of
1
results
1
Indolelactic acid, MedChemExpress
MedChemExpress Indolelactic acid (Indole-3-lactic acid) is a tryptophan (Trp) catabolite in Azotobacter vinelandii cultures. Indolelactic acid has anti-inflammation and potential anti-viral activity.