Learn More
Indolelactic acid, MedChemExpress
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Indolelactic acid (Indole-3-lactic acid) is a tryptophan (Trp) catabolite in Azotobacter vinelandii cultures. Indolelactic acid has anti-inflammation and potential anti-viral activity.
Specifications
Specifications
| Chemical Name or Material | Indolelactic acid |
| CAS | 1821-52-9 |
| Color | Light Brown |
| Purity Grade Notes | Research |
| Molecular Formula | C11H11NO3 |
| Synonym | Indole-3-lactic acid |
| Solubility Information | DMSO : 100 mg/mL (487.31 mM; Need ultrasonic) ∣H2O : 20 mg/mL (97.46 mM; Need ultrasonic) |
| SMILES | OC(C(CC1=CNC2=CC=CC=C12)O)=O |
| Molecular Weight (g/mol) | 205.21 |
| Quantity | 10 mg |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.