accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CCH3CH3(E/Z)

CAS RN 177949-95-0

IUPAC Name: 4,4,5,5-tetramethyl-2-(oct-4-en-4-yl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(oct-4-en-4-yl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 238.18
Molecular Formula: C14H27BO2
InChi Key: MVGKIRIFMYLBJA-UHFFFAOYSA-N
SMILES: CCCC=C(CCC)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results