CAS RN 1703768-74-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Vipivotide tetraxetan Linker, MedChemExpress
MedChemExpress Vipivotide tetraxetan Linker (PSMA-617 Linker) is a nonclaevable peptide linker for synthesis of Vipivotide tetraxetan (PSMA-617).