Learn More
Vipivotide tetraxetan Linker, MedChemExpress
Product Code. 16499865
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
Product Code. Quantity unitSize
16499865 250 mg 250mg
16427836 100 mg 100mg
16413977 50 mg 50mg
3 options
Product Code. 16499865
Brand: Medchem Express HY43869
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress Vipivotide tetraxetan Linker (PSMA-617 Linker) is a nonclaevable peptide linker for synthesis of Vipivotide tetraxetan (PSMA-617).
Vipivotide tetraxetan Linker (PSMA-617 Linker) is a nonclaevable peptide linker for synthesis of Vipivotide tetraxetan (PSMA-617).
Specifications
Chemical Name or Material Vipivotide tetraxetan Linker
CAS 1703768-74-4
Color White
Purity Grade Notes Research
Molecular Formula C33H45N5O9
Quantity 250 mg
Synonym PSMA-617 Linker
Solubility Information DMSO : 270 mg/mL (411.75 mM; Need ultrasonic)
SMILES O=C(O)[C@H](CCCCNC([C@H](CC1=CC=C2C=CC=CC2=C1)NC([C@H]3CC[C@H](CN)CC3)=O)=O)NC(N[C@H](C(O)=O)CCC(O)=O)=O
Molecular Weight (g/mol) 655.74
Formula Weight 655.74
Percent Purity 99.29%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death